Learn More
USA Scientific Inc EASYstrainer Small Filt 40 uM
Catalog No. 502745350
Encompass
EASYstrainer Small Filter 40 uM, for 15 mL standard tubes, green, single packed, 50/cs
Catalog No. 50-274-5350 Supplier USA Scientific Inc Supplier No. 56542140
EASYstrainer Small Filter 40 uM, for 15 mL standard tubes, green, single packed, 50/cs