Learn More
USA Scientific Inc EASYstrainer Small Filt 70 uM
Catalog No. 502745518
Encompass
EASYstrainer Small Filter 70 uM, for 15 mL standard tubes, blue, single packed, 50/cs
Catalog No. 50-274-5518 Supplier USA Scientific Inc Supplier No. 56542170
EASYstrainer Small Filter 70 uM, for 15 mL standard tubes, blue, single packed, 50/cs