Learn More
Sigma Aldrich Fine Chemicals Biosciences Eatons Reagent500ML
Catalog No. 502898524
Encompass
Eaton Reagent contains 7.7wt% phosphorus pentoxide in methanesulfonic acid. It is commonly used as a catalyst and condensing agent.
Catalog No. 50-289-8524 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 380814500ML
Eaton Reagent contains 7.7wt% phosphorus pentoxide in methanesulfonic acid. It is commonly used as a catalyst and condensing agent.