Learn More
Newcomer Supply EasyDip Slide Staining Jar, Blue
SDP
Supplier:  Newcomer Supply 5300B
Encompass
The EasyDip Slide Staining Jar is made of acetal polymer and has an attached lid. The jar uses just 80ml reagent for 12 slides and is resistant to most staining agents including alcohol and xylene. Temperature range: -80°C to +30°C. Dimensions: 2 1/2" x 3" x 3 5/8" (H). The jars may be used individually or linked together and can be used with the EasyDip Slide Staining Rack (sold separately).
Catalog No. NC0634809