Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Eazy Soak, Instrument Cleaner
SDP

Catalog No. 22444305 Shop All Mopec Products
Change view
Click to view available options
pH:
6.5 to 7.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
22-444-305 6.5 to 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-305 Supplier Mopec Supplier No. BE046
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mopec Eazy Soak, Instrument Cleaner digests all tissue, blood and exudative material and is ideal for hard-to-clean instruments such as scissors, forceps, serrated cutting blades and scalpel handles

  • Ideal for overnight soaking
  • Excellent for hard-to-clean instruments
  • No scrubbing or pre-rinsing is needed.
  • Includes liquid dispenser measuring pump
  • Makes 32 gallons of cleaning solution

Specifications

pH 6.5 to 7.5
Appearance White
Container Type Bottle
Content And Storage Store in a dry place between 40 and 100°F
Description Instrument Cleaner
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.