Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EB-42486 | 2390475-81-5 | 99.6% | C22H22N8O | 50 MG
SDP

Supplier:  Medchemexpress LLC HY14264750MG

Encompass_Preferred

EB-42486 is a novel, potent, and highly selective G2019S-LRRK2 inhibitor with an IC50 of less than 0.2 nM. This compound is suitable for research related to Parkinson's disease.

  • Potent and highly selective G2019S-LRRK2 inhibitor
  • Useful in Parkinson's disease research
  • Molecular weight: 414.46
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (125 mg/mL)

Catalog No. 50-003-55503


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.