Learn More
Enzo Life Sciences EB-47. dihydrochloride. dihydrate (5mg)
SDP
Supplier:  Enzo Life Sciences ALX270383M005
Encompass_Preferred
Very potent and water soluble PARP-1 inhibitor (IC50=45nM, 100% inhibition at 200nM). Shows cytoprotective effects against oxidative damage in cells and in in vivo models of reperfusion injury and inflammation. Alternative name: 1-Piperazineacetamide,4-[1-(6-amino-9H-purin-9-yl)-1-deoxy-beta-D-ribofuranuron]-N-(2,3-dihydro-1H-isoindol-4-yl)-1-one . 2HCl . 2H2O. Formula: C24H27N9O6 . 2HCl . 2H2O. MW: 537.5 . 73.0 . 36.0. Purity: ≥95% (HPLC). Appearance: White solid. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C. Handling: Protect from light. Packaged under inert gas.
Catalog No. 50-200-8528