Learn More
Medchemexpress LLC HY-108631 5mg Medchemexpress, EB-47 (dihydrochloride) CAS:1190332-25-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086315MG
Encompass_Preferred
Medchemexpress, HY-108631 5mg EB-47 (dihydrochloride) CAS:1190332-25-2 EB-47 dihydrochloride, a potent and selective PARP-1/ARTD-1 inhibitor with an IC50 value of 45 nM, shows modest potency against ARTD5 with an IC50 value of 410 nM. EB-47 mimics the substrate NAD+ and extends from the nicotinamide to the adenosine subsite. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9640
 Restricted Item