Learn More
Cayman Chemical EB 47hydrochlorIde 10mg
SDP
Catalog No. 502910232
Encompass_Preferred
A PARP1 and TNKS2 inhibitor (IC50 = 45 nM for both); an inhibitor of TNKS1 and PARP10 (IC50s = 410 and 1,179 nM, respectively); reduces infarct volume in a rat model of MCAO occlusion-induced ischemia-reperfusion injury at 10 mg/kg per hour
Catalog No. 50-291-0232 Supplier Cayman Chemical Supplier No. 3468410 MG
A PARP1 and TNKS2 inhibitor (IC50 = 45 nM for both); an inhibitor of TNKS1 and PARP10 (IC50s = 410 and 1,179 nM, respectively); reduces infarct volume in a rat model of MCAO occlusion-induced ischemia-reperfusion injury at 10 mg/kg per hour