Learn More
Medchemexpress LLC Eb-PSMA-617 | 2375590-67-1 | 96.4% | 1938.12 | C88H112N16O28S3 | 1 MG
SDP
Catalog No. 5000201942
Encompass_Preferred
EB-PSMA-617 is an Evans Blue-conjugated ligand that targets prostate-specific membrane antigen (PSMA) to extend blood circulation half-life and enhance tumor uptake for radionuclide targeting and radiotherapy research. It has been evaluated in preclinical models and is supplied as a solid with molecular weight 1938.12, formula C88H112N16O28S3, and purity 96.38%.
  • Conjugated to Evans Blue to prolong blood circulation.
  • Enhances accumulation in PSMA-expressing tumors in preclinical studies.
  • Intended for radionuclide targeting and radiotherapy research applications.
  • Supplied as a 1 mg solid and stored at -20°C to preserve stability.
Catalog No. 50-002-01942 Supplier Medchemexpress LLC Supplier No. HY1636831MG
EB-PSMA-617 is an Evans Blue-conjugated ligand that targets prostate-specific membrane antigen (PSMA) to extend blood circulation half-life and enhance tumor uptake for radionuclide targeting and radiotherapy research. It has been evaluated in preclinical models and is supplied as a solid with molecular weight 1938.12, formula C88H112N16O28S3, and purity 96.38%.
  • Conjugated to Evans Blue to prolong blood circulation.
  • Enhances accumulation in PSMA-expressing tumors in preclinical studies.
  • Intended for radionuclide targeting and radiotherapy research applications.
  • Supplied as a 1 mg solid and stored at -20°C to preserve stability.