Learn More
Medchemexpress LLC Eb-PSMA-617 | 2375590-67-1 | 96.4% | 1938.12 | C88H112N16O28S3 | 2 MG
SDP
Catalog No. 5000203272
Encompass_Preferred
Evans Blue-conjugated PSMA ligand designed to extend circulation half-life and improve prostate tumor uptake for preclinical radiotherapeutic and imaging research.
  • Conjugated to Evans Blue to prolong blood circulation and tumor retention.
  • Improves prostate tumor uptake for targeted radiotherapy studies.
  • Supplied in small research quantities suitable for preclinical experiments.
  • High purity suitable for research use.
  • Characterized by confirmed molecular weight and formula for compound identification.
Catalog No. 50-002-03272 Supplier Medchemexpress LLC Supplier No. HY1636832MG
Evans Blue-conjugated PSMA ligand designed to extend circulation half-life and improve prostate tumor uptake for preclinical radiotherapeutic and imaging research.
  • Conjugated to Evans Blue to prolong blood circulation and tumor retention.
  • Improves prostate tumor uptake for targeted radiotherapy studies.
  • Supplied in small research quantities suitable for preclinical experiments.
  • High purity suitable for research use.
  • Characterized by confirmed molecular weight and formula for compound identification.