Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eb-PSMA-617 | 2375590-67-1 | 96.4% | 1938.12 | C88H112N16O28S3 | 5 MG
SDP

Catalog No. 5000202830
Encompass_Preferred

A PSMA-targeting ligand conjugated to an Evans Blue derivative to extend systemic circulation and enhance tumor uptake for radiotherapeutic use. Preclinical studies report prolonged blood half-life, increased accumulation in PSMA-positive tumors, and tumor regression at reduced radioactivity doses.

  • Molecular weight: 1938.12.
  • Chemical formula: C88H112N16O28S3.
  • Cas number: 2375590-67-1.
  • Purity: 96.4%.
  • Appearance: solid, purple to purplish red.
  • Solubility: soluble in DMSO at 20 mg/mL with ultrasonic assistance.
  • Storage: solid at -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-02830 Supplier Medchemexpress LLC Supplier No. HY1636835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A PSMA-targeting ligand conjugated to an Evans Blue derivative to extend systemic circulation and enhance tumor uptake for radiotherapeutic use. Preclinical studies report prolonged blood half-life, increased accumulation in PSMA-positive tumors, and tumor regression at reduced radioactivity doses.

  • Molecular weight: 1938.12.
  • Chemical formula: C88H112N16O28S3.
  • Cas number: 2375590-67-1.
  • Purity: 96.4%.
  • Appearance: solid, purple to purplish red.
  • Solubility: soluble in DMSO at 20 mg/mL with ultrasonic assistance.
  • Storage: solid at -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.