Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anhydrous Dibasic Calcium Phosphate, Powder, USP, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18600714 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-714 45 kg Fiber Drum
18-600-711 500 g Poly Bottle
18-600-712 2.5 kg Poly Pail
18-600-713 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-714 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C189045KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1890, 7757-93-9, CaHPO4

Spectrum™ Chemical Anhydrous Dibasic Calcium Phosphate, Powder, USP is used as a dietary supplement and as an antacid. It is listed on the FDA′s Generally Recognized As Safe (GRAS) list of substances. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
IUPAC Name calcium hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 45 kg
Packaging Fiber Drum
Percent Purity 98 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.