Learn More
Hettich Centrifuges EBA 200,(8) 13's,16x100,Package
SDP
Catalog No. 502537691 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich EBA 200 BLOOD TUBE PACKAGE 8, (1) Centrifuge, (1) Built-in, Angle Rotor, (8) Adpaters 13x75mm, (8) Adapters 13x100mm, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7691 Supplier Hettich Centrifuges Supplier No. 200BLOOD8
Hettich EBA 200 BLOOD TUBE PACKAGE 8, (1) Centrifuge, (1) Built-in, Angle Rotor, (8) Adpaters 13x75mm, (8) Adapters 13x100mm, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years