Learn More
Hettich Centrifuges EBA 280,(6) 15mL, (3) 50mL,Package
SDP
Catalog No. 502537652 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich EBA 280 CELL CULTURE PACKAGE, (1) Centrifuge, (1) Angle Rotor, (6) Adapters 15mL, (3) Adapters 50mL, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7652 Supplier Hettich Centrifuges Supplier No. 280CELLCULTURE
Add to Cart
Add to Cart
Request a Quote
Hettich EBA 280 CELL CULTURE PACKAGE, (1) Centrifuge, (1) Angle Rotor, (6) Adapters 15mL, (3) Adapters 50mL, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years