Learn More
Hettich Centrifuges EBA 280, (6) Blood Tubes, Package
SDP
Catalog No. 502537655 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich EBA 280 BLOOD TUBE PACKAGE 6, (1) Centrifuge, (1) Horizontal Rotor, (6) Buckets, (6) Adapters 13x75mm, (6) Adapters 16x75mm, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7655 Supplier Hettich Centrifuges Supplier No. 280BLOOD6
Add to Cart
Add to Cart
Request a Quote
Hettich EBA 280 BLOOD TUBE PACKAGE 6, (1) Centrifuge, (1) Horizontal Rotor, (6) Buckets, (6) Adapters 13x75mm, (6) Adapters 16x75mm, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years