Learn More
Hettich Centrifuges EBA 280, (8) Blood Tubes,Package
SDP
Catalog No. 502537653 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich EBA 280 BLOOD TUBE PACKAGE 8, (1) Centrifuge, (1) Horizontal Rotor, (8) Buckets 13x75mm, (8) Buckets 16x75mm, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7653 Supplier Hettich Centrifuges Supplier No. 280BLOOD8
Add to Cart
Add to Cart
Request a Quote
Hettich EBA 280 BLOOD TUBE PACKAGE 8, (1) Centrifuge, (1) Horizontal Rotor, (8) Buckets 13x75mm, (8) Buckets 16x75mm, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years