Learn More
Hettich Centrifuges EBA200S, (8) 0.5mL Pedi, Package
SDP
Catalog No. 502537674 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich EBA 200S PEDIATRIC TUBE PACKAGE 8, (1) Centrifuge, (1) Built-in, Angle Rotor, (8) Adapters 0.5mL, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7674 Supplier Hettich Centrifuges Supplier No. 200SBLOOD8PED
Hettich EBA 200S PEDIATRIC TUBE PACKAGE 8, (1) Centrifuge, (1) Built-in, Angle Rotor, (8) Adapters 0.5mL, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years