Learn More
Apexbio Technology LLC Ebastine 90729-43-4 1g
SDP
Catalog No. 5000094723
Encompass_Preferred
Ebastine (CAS 90729-43-4) is an orally active second-generation histamine H1 receptor antagonist It exerts its pharmacological effect by selectively blocking peripheral H1 receptors thereby inhibiting histamine-mediated allergic responses Ebastine is widely employed in biomedical research to investigate the underlying mechanisms of allergic conditions such as allergic rhinitis including symptoms like nasal pruritus sneezing and rhinorrhea It is also utilized in studies of chronic idiopathic urticaria enabling exploration of disease pathology and symptomatic relief in immune and hypersensitivity models
Catalog No. 50-000-94723 Supplier Apexbio Technology LLC Supplier No. B17401000
Ebastine (CAS 90729-43-4) is an orally active second-generation histamine H1 receptor antagonist It exerts its pharmacological effect by selectively blocking peripheral H1 receptors thereby inhibiting histamine-mediated allergic responses Ebastine is widely employed in biomedical research to investigate the underlying mechanisms of allergic conditions such as allergic rhinitis including symptoms like nasal pruritus sneezing and rhinorrhea It is also utilized in studies of chronic idiopathic urticaria enabling exploration of disease pathology and symptomatic relief in immune and hypersensitivity models