Learn More
Enzo Life Sciences Ebelactone A (1mg). CAS: 76808-16-7
SDP
Supplier:  Enzo Life Sciences ALX270013M001
Encompass_Preferred
Inhibitor of esterases, lipases and N-formylmethionine aminopeptidase. Alternative name: 3,11-Dihydroxy-2,4,6,8,10,12-hexamethyl-9-oxo-6-tetradecenoic 1,3-lactone. Formula: C20H34O4. MW: 338.5. Purity: ≥99%. Solubility: Soluble in methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8368