Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eberconazole nitrate | 130104-32-4 | MFCD00896878 | 392.24 g/mol | C18H15Cl2N3O3 | 1 ML
SDP

Catalog No. 5000240946
Encompass_Preferred

Eberconazole nitrate is an imidazole-class antifungal reagent supplied as a 10 mM solution in DMSO for laboratory research and analytical applications. It is used in vitro for antifungal testing and related studies.

  • Imidazole-class antifungal reagent for in vitro studies.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • Molecular formula C18H15Cl2N3O3; molecular weight 392.24 g/mol.
  • CAS number 130104-32-4.
  • Intended for research and analytical applications; not for human therapeutic use.

Catalog No. 50-002-40946 Supplier Medchemexpress LLC Supplier No. HY106542A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eberconazole nitrate is an imidazole-class antifungal reagent supplied as a 10 mM solution in DMSO for laboratory research and analytical applications. It is used in vitro for antifungal testing and related studies.

  • Imidazole-class antifungal reagent for in vitro studies.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • Molecular formula C18H15Cl2N3O3; molecular weight 392.24 g/mol.
  • CAS number 130104-32-4.
  • Intended for research and analytical applications; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.