Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eberconazole nitrate | 130104-32-4 | MFCD00896878 | 99.8% | 392.24 | C18H15Cl2N3O3 | 25 MG
SDP

Catalog No. 5000240947
Encompass_Preferred

Eberconazole nitrate is a dichlorinated imidazole antifungal compound supplied as an analytical standard for research and analytical applications. It is intended for use in qualitative and quantitative assays, method development, and instrument calibration in chromatographic and mass spectrometric workflows.

  • High purity analytical standard suitable for reference and calibration.
  • Validated by HPLC with purity 99.76%.
  • Molecular formula C18H15Cl2N3O3 and molecular weight 392.24 facilitate compound identification.
  • Available in multiple pack sizes, including 25 mg, for flexible use.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-40947 Supplier Medchemexpress LLC Supplier No. HY106542A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eberconazole nitrate is a dichlorinated imidazole antifungal compound supplied as an analytical standard for research and analytical applications. It is intended for use in qualitative and quantitative assays, method development, and instrument calibration in chromatographic and mass spectrometric workflows.

  • High purity analytical standard suitable for reference and calibration.
  • Validated by HPLC with purity 99.76%.
  • Molecular formula C18H15Cl2N3O3 and molecular weight 392.24 facilitate compound identification.
  • Available in multiple pack sizes, including 25 mg, for flexible use.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.