Learn More
TARGETMOL CHEMICALS INC Eberconazole Nitrate 25MG
SDP
Catalog No. 502876640
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Eberconazole Nitrate (Ebernet) is an azole antifungal agent and broad-spectrum imidazole derivative. Purity 99.53%
Catalog No. 50-287-6640 Supplier TARGETMOL CHEMICALS INC Supplier No. T782425MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Eberconazole Nitrate (Ebernet) is an azole antifungal agent and broad-spectrum imidazole derivative. Purity 99.53%