Learn More
Aviva Systems Biology Corp EBF1 Antibody (ARP39578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EBF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards (50ug) human EBF1.. Alternate names: EBF, COE1, OLF1, O/E-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57090