Learn More
Aviva Systems Biology Corp EBF2 Antibody - middle region (ARP36020_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EBF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EBF2. Alternate names: COE2, EBF-2, FLJ11500, O/E-3, OE-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55973