Learn More
Aviva Systems Biology Corp EBF3 Antibody - middle region (ARP31837_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31837_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EBF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EBF3. Alternate names: COE3, OE-2, EBF-3, O/E-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54336