Learn More
Aviva Systems Biology Corp EBF4 Antibody - C-terminal region (ARP51732_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51732_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EBF4. It was validated on Western Blot. . Alternate names: COE4, KIAA1442, O/E-4, RP5-860F19.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-588