Learn More
Epoch Life Science Inc eBgDy™ Terminator 3.1 Cycle Sequencing Kit, volume 1.0 ml, compatible with BigDye™ Terminator v3.1, applications: de novo sequencing, variant detection, genotyping
SDP
Catalog No. NC2465149
Encompass_Preferred
eBgDy™ Terminator 3.1 Cycle Sequencing Kit, volume 1.0 ml, compatible with BigDye™ Terminator v3.1, applications: de novo sequencing, variant detection, genotyping
Catalog No. NC2465149 Supplier Epoch Life Science Inc Supplier No. C58681.0
eBgDy™ Terminator 3.1 Cycle Sequencing Kit, volume 1.0 ml, compatible with BigDye™ Terminator v3.1, applications: de novo sequencing, variant detection, genotyping