Learn More
Aviva Systems Biology Corp EBI3 Antibody - middle region (ARP42257_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42257_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EBI3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EBI3. Alternate names: IL27B, IL-27BApplicationWB. Species reactivity: Dog, Guinea Pig, Human
Catalog No. 50-014-62938