Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Avidin HRP

Catalog No. 18420089
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
5 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-420-089 5 x 100 μL
50-112-3249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-420-089 Supplier Invitrogen™ Supplier No. 18420089
Add to Cart
Add to Cart

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies.

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.

This product is recommended for use in immunohistochemistry applications.

Conjugate: HRP

Reported Application: Immunohistochemical Staining

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 100 μL
Target Molecule Biotin
Content And Storage Store at 2°C to 8°C
Formulation Aqueous buffer, contains carrier protein/stabilizer, no sodium azide
For Use With (Application) Immunohistochemical Staining
Product Line eBioscience
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.