Learn More
Invitrogen™ eBioscience™ Avidin HRP
Description
Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.
This product is recommended for use in immunohistochemistry applications.
Conjugate: HRP
Reported Application: Immunohistochemical Staining
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C |
| Formulation | Aqueous buffer, contains carrier protein/stabilizer, no sodium azide |
| Form | Liquid |
| For Use With (Application) | Immunohistochemical Staining |
| Product Line | eBioscience |
| Label or Dye | HRP (Horseradish Peroxidase) |
| Target Molecule | Biotin |
| Quantity | 100 μL |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.