Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 506 Conjugate

Catalog No. 69431782
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-431-782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-431-782 Supplier Invitrogen™ Supplier No. 69431782
Add to Cart
Add to Cart

A streptavidin-fluorochrome conjugate for indirect staining protocols to detect biotinylated primary antibodies in flow cytometry

  • eBioscience Streptavidin eFluor 506 is a streptavidin-fluorochrome conjugate for indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. eFluor organic dyes are traditional fluorescent dyes for use in multiple applications, including multicolor flow cytometry and immunofluorescence microscopy.
  • To make fluorophore selection easier, all eFluor organic dyes are named after their emission wavelength, and they are fully compatible with most conventional dyes, fluorescent molecules, eBioscience buffers and fixatives, viability stains, and reagents.
  • The eFluor 506 organic dye is a violet-laser excitable fluorophore that has an emission peak of 506 nm. This dye is optimized for use in flow cytometry and, when combined with all five Super Bright dye antibody conjugates, can provide six detection channels of the violet laser.
  • We recommend using a 510/20 band pass filter or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.
  • Streptavidin eFluor 506 Conjugate has been tested by flow cytometric analysis to detect biotinylated primary antibodies. It can be used ≤ 0.125 μg per test. A test is defined as the amount (μg) of streptavidin fluorochrome conjugate that will stain a cell sample in a final volume of 100 μL.
  • Cell number should be determined empirically but can range from 105 to 108 cells/test. It is recommended that the streptavidin be carefully titrated for optimal performance in the assay of interest.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 506
Quantity 100 μg
Content And Storage Store at 2-8°C.
Shipping Condition Wet Ice
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye eFluor 506

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.