Learn More
Medchemexpress LLC Ebiratide TFA | 99.04% | 996.23 | 1 MG
SDP
Catalog No. 5000400591
Encompass_Preferred
Ebiratide TFA (HOE-427) is an ACTH 4-9 derivative that directly targets the central nervous system, demonstrating memory-enhancing and neuroprotective efficacy. It enhances acetylcholine (ACh) metabolism in rat brain and exhibits anti-amnestic effects in mouse models of memory impairment.
  • Acts directly on the central nervous system
  • Exhibits memory-enhancing efficacy
  • Enhances acetylcholine (ACh) metabolism
  • Exhibits neuroprotective efficacy
  • Exhibits anti-amnestic efficacy
Catalog No. 50-004-00591 Supplier Medchemexpress LLC Supplier No. HY19648B1MG
Ebiratide TFA (HOE-427) is an ACTH 4-9 derivative that directly targets the central nervous system, demonstrating memory-enhancing and neuroprotective efficacy. It enhances acetylcholine (ACh) metabolism in rat brain and exhibits anti-amnestic effects in mouse models of memory impairment.
  • Acts directly on the central nervous system
  • Exhibits memory-enhancing efficacy
  • Enhances acetylcholine (ACh) metabolism
  • Exhibits neuroprotective efficacy
  • Exhibits anti-amnestic efficacy