Learn More
Genscript Corporation eBlot L2 Fast Protein Transfe
Catalog No. 5000059051
Encompass
The eBlot L2 Fast Protein Transfer Device combines the benefits of traditional wet transfer with semi-dry and dry methods supporting transfers of up to 4 mini gels or 2 midi gels in 5 15 minutes for proteins of all sizes
Catalog No. 50-000-59051 Supplier Genscript Corporation Supplier No. L00981
The eBlot L2 Fast Protein Transfer Device combines the benefits of traditional wet transfer with semi-dry and dry methods supporting transfers of up to 4 mini gels or 2 midi gels in 5 15 minutes for proteins of all sizes