Learn More
Lonza Walkersville EBM-2 Endothelial Cell Growth Basal Medium-2, 1 L
Catalog No. NC9993301
Encompass
EBM -2 is an endothelial basal medium sold separately. This can be used with 2 x kits of CC-4176 (Large-vessel endothelial cell growth medium SingleQuots Supplements and Growth Factors) or 2 x CC-4147 (Microvascular endothelial cell growth medium SingleQuots Supplements and Growth Factors).Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed. Any modifications made to the recommended cell systems including the use of alternative media, reagents or protocols, will void cell and media performance guarantees. If you need assistance in selecting the appropriate media, reagents, or protocol, please contact Lonza Scientific Support.Content:1 x Bottle Basal Medium 1 Liter
Catalog No. NC9993301 Supplier Lonza Walkersville Supplier No. 00190860
EBM -2 is an endothelial basal medium sold separately. This can be used with 2 x kits of CC-4176 (Large-vessel endothelial cell growth medium SingleQuots Supplements and Growth Factors) or 2 x CC-4147 (Microvascular endothelial cell growth medium SingleQuots Supplements and Growth Factors).Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed. Any modifications made to the recommended cell systems including the use of alternative media, reagents or protocols, will void cell and media performance guarantees. If you need assistance in selecting the appropriate media, reagents, or protocol, please contact Lonza Scientific Support.Content:1 x Bottle Basal Medium 1 Liter