Learn More
Lonza Walkersville EBM Endothelial Cell Growth Basal Medium, 500 mL
Catalog No. NC9230063
Encompass
EBM is an endothelial basal medium sold separately. This can be used with 1 kit of CC-4133 (endothelial cell growth medium SingleQuots Supplements and Growth Factors for use with certain HUVEC cells).Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed. Any modifications made to the recommended cell systems including the use of alternative media, reagents or protocols, will void cell and media performance guarantees. If you need assistance in selecting the appropriate media, reagents, or protocol, please contact Lonza Scientific Support.Content:1 x Bottle EBM Basal Medium, 500 mL
Catalog No. NC9230063 Supplier Lonza Walkersville Supplier No. CC3121
EBM is an endothelial basal medium sold separately. This can be used with 1 kit of CC-4133 (endothelial cell growth medium SingleQuots Supplements and Growth Factors for use with certain HUVEC cells).Lonza guarantees the performance of Clonetics/Poietics Cells only if appropriate Clonetics/Poietics Media and Reagents are used exclusively and the recommended storage and use protocols are followed. Any modifications made to the recommended cell systems including the use of alternative media, reagents or protocols, will void cell and media performance guarantees. If you need assistance in selecting the appropriate media, reagents, or protocol, please contact Lonza Scientific Support.Content:1 x Bottle EBM Basal Medium, 500 mL