This AC axial fan is designed for air conveyance as a built-in component. It operates at 115 V, providing 520 CFM airflow and a speed of 3200 RPM. Engineered for durability, it features maintenance-free ball bearings with a 40,000-hour service life and is built to industrial standards, ensuring reliable performance in various ambient conditions.
- Operates at 115 volts, 60 hertz
- Provides 520 CFM airflow
- Features a speed of 3200 RPM
- Equipped with maintenance-free ball bearings for extended life
- Offers a degree of protection of IP44
- Includes an internal thermal overload protector for motor safety
- Conforms to EN 60335-1 and CE standards
- Approved by CSA C22.2 No. 77 and UL 1004-3
- Constructed with hot-dip galvanized sheet steel blades and a coated steel guard grille
- Measures 200 mm in size and weighs 3.1 kg
- Manufactured in Germany