This AC centrifugal fan, model R2E225-BE47-09, is designed for efficient air conveyance with a backward-curved impeller. It operates on 230 VAC and is constructed with a durable glass-fiber reinforced plastic impeller, making it suitable for a range of demanding applications requiring reliable air movement.
- Operates on 230 VAC power supply at 60 Hz.
- Delivers an airflow of up to 755 cfm (1285 m³/h) at 0 Pa static pressure.
- Features a fan size of 225 mm in diameter.
- Includes an internally connected thermal overload protector for motor safety.
- Offers an IP44 degree of protection, which is dependent on installation.
- Designed for continuous S1 mode operation.
- Impeller is constructed from PA6 plastic, reinforced with glass-fiber, and has 7 blades.
- Operates effectively in ambient temperatures from -25 °C to +55 °C.