Learn More
MSC EBM Papst Blower, 170 CFM, 115 Volts, Direct Drive
Catalog No. 5000507357
Encompass
This direct drive blower offers 170 CFM airflow and operates at 115 Volts with 3,150 RPM, suitable for various applications requiring efficient air movement.
  • 170 CFM airflow
  • Operates at 115 volts
  • Rotates at 3,150 RPM
  • Direct drive motor
Catalog No. 50-005-07357 Supplier MSC Supplier No. 76940196
This direct drive blower offers 170 CFM airflow and operates at 115 Volts with 3,150 RPM, suitable for various applications requiring efficient air movement.
  • 170 CFM airflow
  • Operates at 115 volts
  • Rotates at 3,150 RPM
  • Direct drive motor