This motorized impeller is designed as a built-in component for efficient air conveyance within a wide range of ambient air pressures and temperatures. It features a robust 1-phase AC motor and a 280 mm diameter impeller, making it suitable for various ventilation applications.
- Conveys air efficiently within specified pressure and temperature ranges.
- Features a 1-phase AC motor for reliable operation.
- Impeller is constructed from durable sheet steel with 11 blades.
- Includes an internal thermal overload protector for enhanced safety.
- Maintenance-free ball bearings ensure a long operational lifespan.
- Offers flexible installation positions, including shaft horizontal or rotor on bottom.
- Compliant with various international safety and performance standards.
- Components are largely recyclable, supporting environmental responsibility.