Learn More
Cayman Chemical Ebola VIrus Envelope Glycopro
SDP
Catalog No. 502945080
Encompass_Preferred
Source: Recombinant Ebola virus (subtype Bundibugyo, strain Uganda 2007) C-Terminal His-tagged envelope glycoprotein GP1 subunit expressed in insect cells • Amino acids: 33-501 • MW: 53.5 kDa
Catalog No. 50-294-5080 Supplier Cayman Chemical Supplier No. 41066100 UG
Source: Recombinant Ebola virus (subtype Bundibugyo, strain Uganda 2007) C-Terminal His-tagged envelope glycoprotein GP1 subunit expressed in insect cells • Amino acids: 33-501 • MW: 53.5 kDa