Learn More
Cayman Chemical Ebola VIrus NucleoproteInsu
SDP
Catalog No. 502945211
Encompass_Preferred
Source: Recombinant Ebola virus N-terminal His-tagged nucleoprotein expressed in E. coli • Amino acids: 630-738 • MW: 14.8 kDa
Catalog No. 50-294-5211 Supplier Cayman Chemical Supplier No. 41064100 UG
Source: Recombinant Ebola virus N-terminal His-tagged nucleoprotein expressed in E. coli • Amino acids: 630-738 • MW: 14.8 kDa