Learn More
Cayman Chemical Ebola VIrus VP40subTYP Sud
SDP
Catalog No. 502945173
Encompass_Preferred
Source: Recombinant Ebola virus N-terminal His-tagged VP40 expressed in E. coli • Amino acids: 31-326 • MW: 34.5 kDa
Catalog No. 50-294-5173 Supplier Cayman Chemical Supplier No. 41065100 UG
Source: Recombinant Ebola virus N-terminal His-tagged VP40 expressed in E. coli • Amino acids: 31-326 • MW: 34.5 kDa