Learn More
Medchemexpress LLC Ebopiprant | 2005486-31-5 | 98.7% | 599.74 | 100 MG
SDP
Catalog No. 5000435733
Encompass_Preferred
Ebopiprant is an oral and selective prostaglandin F2α (PGF2α) receptor antagonist. It is characterized by its high affinity for FP receptors.
  • Oral and selective prostaglandin F2α (PGF2α) receptor antagonist
  • Ki values of 1 nM for human FP receptors and 26 nM for rat FP receptors
Catalog No. 50-004-35733 Supplier Medchemexpress LLC Supplier No. HY112284100MG
Ebopiprant is an oral and selective prostaglandin F2α (PGF2α) receptor antagonist. It is characterized by its high affinity for FP receptors.
  • Oral and selective prostaglandin F2α (PGF2α) receptor antagonist
  • Ki values of 1 nM for human FP receptors and 26 nM for rat FP receptors