Learn More
Sino Biological EBOV EBOV-G MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40094R10620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Sudan ebolavirus Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP1 (mucin domain deleted) (rSudan ebolavirus Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP1 (mucin domain deleted); Catalog#40094-V08H; Q7T9D9; Met1-Asp320).
Catalog No. 50-237-3204
 Restricted Item