Learn More
Sino Biological EBOV EBOV-G MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40094R30120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Sudan ebolavirus EBOV-G / Glycoprotein (rSudan ebolavirus EBOV-G / Glycoprotein; Catalog#40094-V08H; Q7T9D9; Met1-Asp320).
Catalog No. 50-237-1463