Learn More
Sino Biological Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP1 (mucin domain deleted) Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40094T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP1 (mucin domain deleted) Protein (Catalog#40094-V08H; Q7T9D9; Met1-Asp320).
Catalog No. 50-240-8949