Learn More
Sino Biological Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40094T46100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP ( Catalog#40094-V08B1; YP_138523.1; Met1-Asn637). Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP specific IgG was purified by Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP affinity chromatography.
Catalog No. 50-240-8941