Learn More
Sino Biological EBOV EBOV-G PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11503R014100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Sudan ebolavirus EBOV-G / Glycoprotein (Catalog#40094-V08B1; YP_138523.1; Met1-Asn637). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4141