Learn More
Sino Biological Ebola virus Nucleoprotein Antibody, Mouse MAb
Supplier:  Sino Biological 40443MM0920
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant EBOV Ebola virus Nucleoprotein ( Catalog#40443-V07E1; AHX24646.1; His630-Gln739). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8699